Assay Details
| Gene Symbol: FSCN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018063601.1 | XP_017919090.1 | 1 - 2 | 1033 | 64 | |
| XM_018063602.1 | XP_017919091.1 | 1 - 2 | 1033 | 64 | ||
More Information
Set Membership: |
|

FSCN2
Genomic Map