Assay Details
| Gene Symbol: RHEBL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005680005.2 | XP_005680062.1 | 6 - 7 | 619 | 61 | |
| GenBank mRNA | KA315555.1 | - | 4 - 5 | 229 | 61 | |
More Information
Set Membership: |
|

RHEBL1
Genomic Map