Assay Details
| Gene Symbol: NACA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005680328.3 | XP_005680385.1 | 4 - 5 | 349 | 62 | |
| XM_005680329.3 | XP_005680386.1 | 5 - 6 | 471 | 62 | ||
| XM_013963935.1 | XP_013819389.1 | 4 - 5 | 344 | 62 | ||
| GenBank mRNA | JO445028.2 | - | 4 - 5 | 528 | 62 | |
| JO633862.2 | - | 4 - 5 | 538 | 62 | ||
More Information
Set Membership: |
|

NACA
Genomic Map