Assay Details
| Gene Symbol: RIDA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285713.1 | NP_001272642.1 | 2 - 3 | 280 | 91 | |
| GenBank mRNA | AF000167.1 | - | 2 - 3 | 280 | 91 | |
| JO333451.2 | - | 4 - 5 | 729 | 91 | ||
| JO439303.2 | - | 4 - 5 | 815 | 91 | ||
More Information
Set Membership: |
|

RIDA
Genomic Map