Assay Details
| Gene Symbol: PSMB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005677722.3 | XP_005677779.2 | 4 - 5 | 624 | 61 | |
| GenBank mRNA | JO580356.1 | - | 5 - 6 | 885 | 61 | |
More Information
Set Membership: |
|

PSMB4
Genomic Map