Assay Details
| Gene Symbol: SRSF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005686512.3 | XP_005686569.1 | 3 - 4 | 617 | 63 | |
| XM_005686513.3 | XP_005686570.1 | 3 - 4 | 617 | 63 | ||
| XM_005686514.3 | XP_005686571.1 | 3 - 4 | 617 | 63 | ||
| GenBank mRNA | KA347158.1 | - | 3 - 4 | 500 | 63 | |
More Information
Set Membership: |
|

SRSF7
Genomic Map