Assay Details
| Gene Symbol: TPPP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013966938.2 | XP_013822392.2 | 3 - 4 | 394 | 54 | |
| XM_018054526.1 | XP_017910015.1 | 4 - 5 | 776 | 54 | ||
| XM_018054527.1 | XP_017910016.1 | 4 - 5 | 562 | 54 | ||
| XM_018054528.1 | XP_017910017.1 | 4 - 5 | 713 | 54 | ||
| XM_018054529.1 | XP_017910018.1 | 4 - 5 | 697 | 54 | ||
| XM_018054530.1 | XP_017910019.1 | 3 - 4 | 628 | 54 | ||
| GenBank mRNA | JO413718.2 | - | 1 - 2 | 294 | 54 | |
More Information
Set Membership: |
|

TPPP2
Genomic Map