Assay Details
| Gene Symbol: VAMP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018055378.1 | XP_017910867.1 | 2 - 3 | 408 | 64 | |
| GenBank mRNA | JO447211.2 | - | 1 - 2 | 444 | 64 | |
| JO580971.1 | - | 1 - 2 | 504 | 64 | ||
More Information
Set Membership: |
|

VAMP8
Genomic Map