Assay Details
| Gene Symbol: PERP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005684810.3 | XP_005684867.1 | 1 - 2 | 417 | 66 | |
| GenBank mRNA | KA332099.1 | - | 1 - 2 | 509 | 66 | |
More Information
Set Membership: |
|

PERP
Genomic Map