Assay Details
| Gene Symbol: POLR1E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005683878.3 | XP_005683935.2 | 2 - 3 | 256 | 61 | |
| XM_018052186.1 | XP_017907675.1 | 2 - 3 | 188 | 61 | ||
| GenBank mRNA | KA316473.1 | - | 10 - 11 | 1342 | 61 | |
More Information
Set Membership: |
|

POLR1E
Genomic Map