Assay Details
| Gene Symbol: VPS29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005691431.2 | XP_005691488.1 | 3 - 4 | 302 | 72 | |
| XM_005691432.3 | XP_005691489.1 | 2 - 3 | 297 | 72 | ||
| GenBank mRNA | KA348181.1 | - | 391 | 72 | ||
More Information
Set Membership: |
|

VPS29
Genomic Map