Assay Details
| Gene Symbol: BDH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005681356.3 | XP_005681413.1 | 6 - 7 | 534 | 61 | |
| XM_005681357.3 | XP_005681414.1 | 6 - 7 | 618 | 61 | ||
| XM_005681358.3 | XP_005681415.1 | 7 - 8 | 702 | 61 | ||
| XM_018049253.1 | XP_017904742.1 | 7 - 8 | 827 | 61 | ||
| XM_018049254.1 | XP_017904743.1 | 7 - 8 | 787 | 61 | ||
| XM_018049255.1 | XP_017904744.1 | 7 - 8 | 736 | 61 | ||
More Information
Set Membership: |
|

BDH2
Genomic Map