Assay Details
| Gene Symbol: MRPL16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018059017.1 | XP_017914506.1 | 2 - 3 | 223 | 70 | |
| GenBank mRNA | JO411465.2 | - | 2 - 3 | 1026 | 70 | |
| JO635871.2 | - | 2 - 3 | 835 | 70 | ||
More Information
Set Membership: |
|

MRPL16
Genomic Map