Assay Details
| Gene Symbol: PDZD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018059674.1 | XP_017915163.1 | 5 - 6 | 605 | 60 | |
| XM_018059675.1 | XP_017915164.1 | 5 - 6 | 479 | 60 | ||
More Information
Set Membership: |
|

PDZD3
Genomic Map