Assay Details
| Gene Symbol: STMN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005676827.3 | XP_005676884.1 | 4 - 5 | 463 | 70 | |
| XM_018056181.1 | XP_017911670.1 | 4 - 5 | 551 | 70 | ||
| GenBank mRNA | JO339694.2 | - | 1 - 2 | 545 | 70 | |
| KA335298.1 | - | 1 - 2 | 520 | 70 | ||
More Information
Set Membership: |
|

STMN1
Genomic Map