Assay Details
| Gene Symbol: MLX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013972164.2 | XP_013827618.1 | 7 - 8 | 897 | 71 | |
| XM_018065128.1 | XP_017920617.1 | 7 - 8 | 756 | 71 | ||
| XM_018065129.1 | XP_017920618.1 | 6 - 7 | 666 | 71 | ||
More Information
Set Membership: |
|

MLX
Genomic Map