Assay Details
| Gene Symbol: TCP11L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018059272.1 | XP_017914761.1 | 6 - 7 | 1105 | 63 | |
| XM_018059273.1 | XP_017914762.1 | 6 - 7 | 959 | 63 | ||
| XM_018059274.1 | XP_017914763.1 | 6 - 7 | 963 | 63 | ||
| XM_018059275.1 | XP_017914764.1 | 6 - 7 | 1072 | 63 | ||
| XM_018059276.1 | XP_017914765.1 | 6 - 7 | 877 | 63 | ||
| XM_018059277.1 | XP_017914766.1 | 5 - 6 | 913 | 63 | ||
| XM_018059278.1 | XP_017914767.1 | 5 - 6 | 714 | 63 | ||
| XM_018059279.1 | XP_017914768.1 | 6 - 7 | 798 | 63 | ||
| GenBank mRNA | JO631003.2 | - | 6 - 7 | 1138 | 63 | |
More Information
Set Membership: |
|

TCP11L1
Genomic Map