Assay Details
| Gene Symbol: LOC102185066 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005680325.3 | XP_005680382.2 | 3 - 4 | 685 | 70 | |
| XM_005680326.3 | XP_005680383.2 | 3 - 4 | 675 | 70 | ||
| GenBank mRNA | JO420208.2 | - | 2 - 3 | 949 | 70 | |
More Information
Set Membership: |
|

LOC102185066
Genomic Map