Assay Details
| Gene Symbol: BPGM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005679489.3 | XP_005679546.1 | 2 - 3 | 793 | 72 | |
| XM_018046912.1 | XP_017902401.1 | 3 - 4 | 1025 | 72 | ||
| XM_018046913.1 | XP_017902402.1 | 3 - 4 | 1002 | 72 | ||
| XM_018046914.1 | XP_017902403.1 | 3 - 4 | 925 | 72 | ||
| GenBank mRNA | JO360690.2 | - | 2 - 3 | 773 | 72 | |
More Information
Set Membership: |
|

BPGM
Genomic Map