Assay Details
| Gene Symbol: SLC16A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013962525.2 | XP_013817979.1 | 3 - 4 | 503 | 89 | |
| XM_018045927.1 | XP_017901416.1 | 3 - 4 | 564 | 89 | ||
| XM_018045928.1 | XP_017901417.1 | 4 - 5 | 753 | 89 | ||
| GenBank mRNA | AB231662.1 | - | 1 - 2 | 118 | 89 | |
More Information
Set Membership: |
|

SLC16A1
Genomic Map