Assay Details
| Gene Symbol: ZBTB16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001314273.1 | NP_001301202.1 | 3 - 4 | 1611 | 66 | |
| XM_018058854.1 | XP_017914343.1 | 4 - 5 | 2116 | 66 | ||
| XM_018058856.1 | XP_017914345.1 | 4 - 5 | 1835 | 66 | ||
| XM_018058857.1 | XP_017914346.1 | 3 - 4 | 1667 | 66 | ||
| GenBank mRNA | JX047313.1 | - | 3 - 4 | 1611 | 66 | |
More Information
Set Membership: |
|

ZBTB16
Genomic Map