Assay Details
| Gene Symbol: PKIA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005689114.3 | XP_005689171.1 | 2 - 3 | 270 | 87 | |
| XM_005689116.3 | XP_005689173.1 | 3 - 4 | 454 | 87 | ||
| XM_018058384.1 | XP_017913873.1 | 4 - 5 | 573 | 87 | ||
| XM_018058385.1 | XP_017913874.1 | 2 - 3 | 327 | 87 | ||
More Information
Set Membership: |
|

PKIA
Genomic Map