Assay Details
| Gene Symbol: SRPRA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005699612.3 | XP_005699669.2 | 9 - 10 | 1319 | 70 | |
| GenBank mRNA | JO426580.1 | - | 5 - 6 | 1812 | 70 | |
| JO496168.2 | - | 2055 | 70 | |||
| JO643572.2 | - | 5 - 6 | 1736 | 70 | ||
More Information
Set Membership: |
|

SRPRA
Genomic Map