Assay Details
| Gene Symbol: EMP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018040779.1 | XP_017896268.1 | 4 - 5 | 495 | 85 | |
| GenBank mRNA | JO421992.2 | - | 4 - 5 | 496 | 85 | |
More Information
Set Membership: |
|

EMP2
Genomic Map