Assay Details
| Gene Symbol: PLPP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005678357.3 | XP_005678414.1 | 3 - 4 | 1159 | 60 | |
| XM_018044641.1 | XP_017900130.1 | 3 - 4 | 1166 | 60 | ||
| GenBank mRNA | KA352846.1 | - | 3 - 4 | 524 | 60 | |
More Information
Set Membership: |
|

PLPP3
Genomic Map