Assay Details
| Gene Symbol: WASHC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005680525.3 | XP_005680582.1 | 6 - 7 | 665 | 91 | |
| XM_005680526.3 | XP_005680583.1 | 6 - 7 | 662 | 91 | ||
| GenBank mRNA | JO628235.2 | - | 2 - 3 | 117 | 91 | |
More Information
Set Membership: |
|

WASHC3
Genomic Map