Assay Details
| Gene Symbol: PAEP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285539.1 | NP_001272468.1 | 4 - 5 | 458 | 103 | |
| XM_018054687.1 | XP_017910176.1 | 4 - 5 | 634 | 103 | ||
| XM_018054688.1 | XP_017910177.1 | 4 - 5 | 634 | 103 | ||
| XM_018054689.1 | XP_017910178.1 | 4 - 5 | 634 | 103 | ||
| GenBank mRNA | EF564255.1 | - | 4 - 5 | 448 | 103 | |
| JO517063.2 | - | 3 - 4 | 233 | 103 | ||
| X58471.1 | - | 4 - 5 | 430 | 103 | ||
| Z19569.1 | - | 4 - 5 | 458 | 103 | ||
More Information
Set Membership: |
|

PAEP
Genomic Map