Assay Details
| Gene Symbol: DHRS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005690659.3 | XP_005690716.1 | 3 - 4 | 1471 | 65 | |
| GenBank mRNA | JO432803.2 | - | 3 - 4 | 814 | 65 | |
More Information
Set Membership: |
|

DHRS3
Genomic Map