Assay Details
| Gene Symbol: C10H14orf93 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018054581.1 | XP_017910070.1 | 6 - 7 | 1701 | 83 | |
| XM_018054582.1 | XP_017910071.1 | 6 - 7 | 1514 | 83 | ||
| XM_018054583.1 | XP_017910072.1 | 6 - 7 | 1459 | 83 | ||
| XM_018054584.1 | XP_017910073.1 | 6 - 7 | 1527 | 83 | ||
| GenBank mRNA | JO592208.2 | - | 5 - 6 | 728 | 83 | |
More Information
Set Membership: |
|

C10H14orf93
Genomic Map