Assay Details
| Gene Symbol: EMC8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018061816.1 | XP_017917305.1 | 4 - 5 | 800 | 55 | |
| GenBank mRNA | JT745422.1 | - | 2 - 3 | 544 | 55 | |
More Information
Set Membership: |
|

EMC8
Genomic Map