Assay Details
| Gene Symbol: SRP54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005695231.3 | XP_005695288.1 | 5 - 6 | 621 | 72 | |
| XM_013973098.2 | XP_013828552.1 | 6 - 7 | 744 | 72 | ||
| GenBank mRNA | JT762390.1 | - | 11 - 12 | 1709 | 72 | |
More Information
Set Membership: |
|

SRP54
Genomic Map