Assay Details
| Gene Symbol: PSMB7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005687126.3 | XP_005687183.1 | 4 - 5 | 455 | 63 | |
| GenBank mRNA | JO581286.1 | - | 4 - 5 | 625 | 63 | |
More Information
Set Membership: |
|

PSMB7
Genomic Map