Assay Details
| Gene Symbol: SPAG7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005693453.3 | XP_005693510.1 | 6 - 7 | 1522 | 61 | |
| GenBank mRNA | JO411209.2 | - | 6 - 7 | 716 | 61 | |
More Information
Set Membership: |
|

SPAG7
Genomic Map