Assay Details
Gene Symbol: RNF141 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005689690.3 | XP_005689747.1 | 3 - 4 | 565 | 64 | |
XM_005689691.3 | XP_005689748.1 | 3 - 4 | 648 | 64 | ||
GenBank mRNA | JO431498.2 | - | 3 - 4 | 1492 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
