Assay Details
| Gene Symbol: LOC102180196 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018043080.1 | XP_017898569.1 | 3 - 4 | 1240 | 64 | |
| XM_018043081.1 | XP_017898570.1 | 3 - 4 | 1282 | 64 | ||
| GenBank mRNA | JT754481.1 | - | 11 - 12 | 3895 | 64 | |
More Information
Set Membership: |
|

LOC102180196
Genomic Map