Assay Details
Gene Symbol: PSMA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285698.1 | NP_001272627.1 | 6 - 7 | 716 | 123 | |
GenBank mRNA | FJ594753.1 | - | 6 - 7 | 716 | 123 | |
JO580480.1 | - | 1 - 2 | 295 | 123 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
