Assay Details
| Gene Symbol: ACYP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005686091.3 | XP_005686148.2 | 2 - 3 | 382 | 74 | |
| XM_005686092.3 | XP_005686149.1 | 2 - 3 | 242 | 74 | ||
| XM_005686093.3 | XP_005686150.1 | 2 - 3 | 300 | 74 | ||
| GenBank mRNA | JO451008.2 | - | 2 - 3 | 334 | 74 | |
| JO600449.2 | - | 2 - 3 | 236 | 74 | ||
More Information
Set Membership: |
|

ACYP1
Genomic Map