Assay Details
| Gene Symbol: ACBD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005693947.3 | XP_005694004.1 | 4 - 5 | 1846 | 54 | |
| XM_005693949.3 | XP_005694006.1 | 4 - 5 | 1841 | 54 | ||
| XM_005693950.3 | XP_005694007.1 | 4 - 5 | 1839 | 54 | ||
| GenBank mRNA | JO430332.2 | - | 7 - 8 | 1329 | 54 | |
More Information
Set Membership: |
|

ACBD4
Genomic Map