Assay Details
Gene Symbol: C8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005678355.3 | XP_005678412.1 | 3 - 4 | 423 | 77 | |
GenBank mRNA | JO419668.2 | - | 3 - 4 | 466 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
