Assay Details
| Gene Symbol: NONO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005700665.3 | XP_005700722.1 | 2 - 3 | 417 | 94 | |
| XM_013976413.2 | XP_013831867.1 | 2 - 3 | 291 | 94 | ||
| GenBank mRNA | JP351039.2 | - | 9 - 10 | 1876 | 94 | |
More Information
Set Membership: |
|

NONO
Genomic Map