Assay Details
| Gene Symbol: C5H12orf57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005680927.3 | XP_005680984.2 | 2 - 3 | 544 | 84 | |
| GenBank mRNA | JO581239.1 | - | 1 - 2 | 298 | 84 | |
| JO632188.2 | - | 2 - 3 | 523 | 84 | ||
More Information
Set Membership: |
|

C5H12orf57
Genomic Map