Assay Details
Gene Symbol: WDR53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005675069.2 | XP_005675126.2 | 4 - 5 | 670 | 62 | |
XM_005675070.2 | XP_005675127.2 | 3 - 4 | 580 | 62 | ||
XM_013963522.2 | XP_013818976.2 | 2 - 3 | 558 | 62 | ||
XM_018046257.1 | XP_017901746.1 | 3 - 4 | 586 | 62 | ||
GenBank mRNA | JO361033.2 | - | 1 - 2 | 432 | 62 | |
JO386985.2 | - | 1 - 2 | 100 | 62 | ||
JO417717.2 | - | 1 - 2 | 623 | 62 | ||
JO438064.2 | - | 1 - 2 | 531 | 62 | ||
JO442183.2 | - | 2 - 3 | 298 | 62 | ||
JO568431.2 | - | 1 - 2 | 165 | 62 | ||
JO571962.2 | - | 1 - 2 | 165 | 62 | ||
JO589063.2 | - | 1 - 2 | 629 | 62 | ||
KA347108.1 | - | 1 - 2 | 531 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
