Assay Details
| Gene Symbol: TBPL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005684780.3 | XP_005684837.2 | 4 - 5 | 576 | 83 | |
| GenBank mRNA | KA350925.1 | - | 3 - 4 | 681 | 83 | |
More Information
Set Membership: |
|

TBPL1
Genomic Map