Assay Details
| Gene Symbol: SLC39A11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005694468.3 | XP_005694525.2 | 6 - 7 | 741 | 58 | |
| XM_005694469.3 | XP_005694526.2 | 6 - 7 | 720 | 58 | ||
| XM_018063902.1 | XP_017919391.1 | 6 - 7 | 796 | 58 | ||
| XM_018063903.1 | XP_017919392.1 | 6 - 7 | 775 | 58 | ||
More Information
Set Membership: |
|

SLC39A11
Genomic Map