Assay Details
| Gene Symbol: ASB17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005678234.1 | XP_005678291.1 | 2 - 3 | 810 | 95 | |
| XM_013962661.1 | XP_013818115.1 | 2 - 3 | 635 | 95 | ||
More Information
Set Membership: |
|

ASB17
Genomic Map