Assay Details
Gene Symbol: CSNK2A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018057661.1 | XP_017913150.1 | 11 - 12 | 1251 | 63 | |
XM_018057662.1 | XP_017913151.1 | 11 - 12 | 1201 | 63 | ||
GenBank mRNA | KA338656.1 | - | 11 - 12 | 1211 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
