Assay Details
| Gene Symbol: CCNB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005694606.3 | XP_005694663.1 | 2 - 3 | 387 | 95 | |
| GenBank mRNA | JO611848.2 | - | 7 - 8 | 1272 | 95 | |
| KA307994.1 | - | 2 - 3 | 314 | 95 | ||
More Information
Set Membership: |
|

CCNB1
Genomic Map