Assay Details
| Gene Symbol: GATM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018054151.1 | XP_017909640.1 | 6 - 7 | 1151 | 74 | |
| GenBank mRNA | JO632708.2 | - | 3 - 4 | 681 | 74 | |
More Information
Set Membership: |
|

GATM
Genomic Map