Assay Details
| Gene Symbol: PTDSS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018058253.1 | XP_017913742.1 | 9 - 10 | 1181 | 60 | |
| XM_018058254.1 | XP_017913743.1 | 8 - 9 | 876 | 60 | ||
| GenBank mRNA | JT683234.1 | - | 7 - 8 | 1116 | 60 | |
More Information
Set Membership: |
|

PTDSS1
Genomic Map