Assay Details
| Gene Symbol: MALL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005686240.3 | XP_005686297.1 | 1 - 2 | 175 | 66 | |
| GenBank mRNA | JO429140.2 | - | 3 - 4 | 1863 | 66 | |
More Information
Set Membership: |
|

MALL
Genomic Map